Merck
A recessive Trim2 mutation causes an axonal neuropathy in mice.
Neurobiology of disease (2020-03-25)
Jian J Li, Nicolas Sarute, Eunjoo Lancaster, Guliz Otkiran-Clare, Bani Medegan Fagla, Susan R Ross, Steven S Scherer
ABSTRAKT
We analyzed Trim2A/A mice, generated by CRISPR-Cas9, which have a recessive, null mutation of Trim2. Trim2A/A mice develop ataxia that is associated with a severe loss of cerebellar Purkinje cells and a peripheral neuropathy. Myelinated axons in the CNS, including those in the deep cerebellar nuclei, have focal enlargements that contain mitochondria and neurofilaments. In the PNS, there is a loss of myelinated axons, particularly in the most distal nerves. The pathologically affected neuronal populations - primary sensory and motor neurons as well as cerebellar Purkinje cells - express TRIM2, suggesting that loss of TRIM2 in these neurons results in cell autonomous effects on their axons. In contrast, these pathological findings were not found in a second strain of Trim2 mutant mice (Trim2C/C), which has a partial deletion in the RING domain that is needed for ubiquitin ligase activity. Both the Trim2Aand the Trim2C alleles encode mutant TRIM2 proteins with reduced ubiquitination activity. In sum, Trim2A/A mice are a genetically authentic animal model of a recessive axonal neuropathy of humans, apparently for a function that does not depend on the ubiquitin ligase activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Calbindin-D-28K (EG-20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-TRIM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution