Merck
Oncofetal protein IGF2BP1 regulates IQGAP3 expression to maintain stem cell potential in cancer.
iScience (2022-10-12)
Khine Myint, Linda Shyue Huey Chuang, Yu Xuan Teh, Nur Astiana Mawan, Edward Jizhong Shi, Michelle Meng Huang Mok, Napat Nuttonmanit, Junichi Matsuo, Ying Li, Henry Yang, Atsushi Okabe, Atsushi Kaneda, Motomi Osato, Jimmy Bok-Yan So, Wei Peng Yong, Patrick Tan, Khay Guan Yeoh, Yoshiaki Ito
ABSTRAKT
We reported earlier that IQGAP3 is an important stem cell factor in rapidly proliferating isthmus stem cells in the stomach and that IQGAP3 expression is robustly induced in terminally differentiated chief cells and de-differentiated cells following tissue damage. The elevated IQGAP3 expression in cancer and its association with metastasis suggest a fundamental role for IQGAP3 in proliferating cancer stem cells. What causes IQGAP3 upregulation in cancer is unclear. Here, we show that IGF2BP1 and IQGAP3 expression levels are highest in the blastocyst, with both decreasing during adulthood. This suggests that IQGAP3, like IGF2BP1, is an early developmental gene that is aberrantly upregulated upon re-expression of IGF2BP1 during carcinogenesis. IGF2BP1 binds and stabilizes m6A-modified IQGAP3 transcripts. Downstream targets of IGF2BP1, namely SRF and FOXM1, also upregulate IQGAP3 expression. These multiple layers of IQGAP3 regulation, which may safeguard against inappropriate stem cell proliferation, present additional drug targets to inhibit IQGAP3-driven malignant growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-N6-methyladenosine Antibody (m6A), clone 17-3-4-1, clone 17-3-4-1, from mouse
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%