Merck
Effects of maternal choline supplementation on performance and immunity of progeny from birth to weaning.
Journal of dairy science (2022-10-15)
M G Zenobi, J M Bollatti, A M Lopez, B A Barton, C L Hixson, F P Maunsell, W W Thatcher, K Miller-Cushon, J E P Santos, C R Staples, C D Nelson
ABSTRAKT
The objectives were to investigate whether supplementation with rumen-protected choline (RPC) during late pregnancy in Holstein cows affects offspring immunity and growth, and whether effects are utero-placental, colostrum dependent, or both. A total of 105 multiparous Holstein cows were assigned randomly to a prepartum diet (1.54 Mcal of NEL/kg of DM, and 15.8% CP) without (control) or with added RPC (12.9 g/d of choline ion). Calves (n = 111) were blocked by sex and assigned randomly to colostrum from control cows or colostrum from RPC cows, resulting in 4 treatments in a 2 × 2 factorial arrangement: (1) calves born and fed colostrum from non-supplemented dams (NN; n = 33); (2) calves from non-supplemented dams and fed colostrum from RPC-fed cows (NC; n = 25); (3) calves from RPC-supplemented dams and colostrum from non-supplemented cows (CN; n = 28); and (4) calves from RPC-supplemented dams and colostrum from RPC-fed cows (CC; n = 25). Growth, intakes, and immunity of females were evaluated up to 56 d of age. Growth and intake of male calves was evaluated up to 35 d of age, and physiological and immune responses to intravenous LPS challenge were evaluated from 21 to 35 d of age. Effects of prenatal and colostrum treatments and interactions between treatments were analyzed using mixed models. Calves fed colostrum from RPC-supplemented dams had a 17.4% increase in apparent efficiency of absorption of IgG compared with calves fed colostrum from control dams (27.4 vs. 23.3%). Incidence of fever in the first 21 d of age tended to be less in females born from RPC-supplemented dams compared with females born from control dams (31 vs. 58%). Prenatal RPC females had increased hematocrit and concentrations of red blood cells, leukocytes, neutrophils, and lymphocytes in blood compared with prenatal females born from control dams. Compared with prenatal control females, prenatal RPC females had greater intake of milk replacer (704 vs. 748 ± 9.9 g/d) and starter (45.4 vs. 60.2 ± 5.9 g/d) during the first 21 d of age. In male calves, mean intake of DM was greater (1,074 vs. 976 ± 45 g/d) after the LPS challenge (0 to 8 d) by calves born from dams fed RPC compared with males born from control dams. Calves born from RPC-fed dams also had lower mean rectal temperature (39.0 vs. 39.2°C) and mean respiration rate (35.6 vs. 39.3 breaths/min) compared with males born from control dams. Moreover, serum concentrations of metabolites (i.e., β-hydroxybutyric acid, fatty acids, and glucose), cytokines (i.e., tumor necrosis factor-α) and acute phase proteins (i.e., serum amyloid A) were consistent with less-severe inflammatory response to LPS in males born from dams fed RPC compared with control. Source of colostrum and interaction between prenatal and colostrum treatments had minimal effects on calf responses to LPS. Overall, maternal RPC supplementation during late gestation suggests a positive effect on immunity, in that colostrum from RPC-fed dams increased efficiency of IgG absorption and maternal supplementation with RPC during late gestation, regardless of colostrum source, attenuated responses to LPS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-P, lyophilized powder (contains ~50% buffer salts as sodium phosphate and NaCl), BioReagent, suitable for cell culture