Merck
SPIN90 Deficiency Ameliorates Amyloid β Accumulation by Regulating APP Trafficking in AD Model Mice.
International journal of molecular sciences (2022-09-24)
Youngsoo Oh, Wongyoung Lee, So Hee Kim, Sooji Lee, Byeong C Kim, Kun Ho Lee, Sung Hyun Kim, Woo Keun Song
ABSTRAKT
Alzheimer's disease (AD), a common form of dementia, is caused in part by the aggregation and accumulation in the brain of amyloid β (Aβ), a product of the proteolytic cleavage of amyloid precursor protein (APP) in endosomes. Trafficking of APP, such as surface-intracellular recycling, is an early critical step required for Aβ generation. Less is known, however, about the molecular mechanism regulating APP trafficking. This study investigated the mechanism by which SPIN90, along with Rab11, modulates APP trafficking, Aβ motility and accumulation, and synaptic functionality. Brain Aβ deposition was lower in the progeny of 5xFAD-SPIN90KO mice than in 5xFAD-SPIN90WT mice. Analysis of APP distribution and trafficking showed that the surface fraction of APP was locally distinct in axons and dendrites, with these distributions differing significantly in 5xFAD-SPIN90WT and 5xFAD-SPIN90KO mice, and that neural activity-driven APP trafficking to the surface and intracellular recycling were more actively mobilized in 5xFAD-SPIN90KO neurons. In addition, SPIN90 was found to be cotrafficked with APP via axons, with ablation of SPIN90 reducing the intracellular accumulation of APP in axons. Finally, synaptic transmission was restored over time in 5xFAD-SPIN90KO but not in 5xFAD-SPIN90WT neurons, suggesting SPIN90 is implicated in Aβ production through the regulation of APP trafficking.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-BACE 1, N-Terminus (46-62) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse