Merck
c-Myc target genes involved in cell growth, apoptosis, and metabolism.
Molecular and cellular biology (1998-12-22)
C V Dang
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
c-Myc (EP121) Rabbit Monoclonal Primary Antibody