Merck
In vitro vascularization of human connective microtissues.
Methods in molecular medicine (2007-12-19)
Jens M Kelm, Wolfgang Moritz, Doerthe Schmidt, Simon P Hoerstrup, Martin Fussenegger
ABSTRAKT
Vascularization is one of the most central processes enabling multicellular life. Owing to the complexity of vascularization regulatory networks, minor control imbalances often have severe pathologic consequences ranging from ischemic diseases to cancer. Tissue engineers are immediately confronted with vascularization as artificial tissues of a clinically relevant size require a vascular system to ensure vital physiologic logistics throughout the entire tissue and to enable rapid connection to the host vasculature following implantation. Using human umbilical vein endothelial cells (HUVECs) coated onto a human aortic fibroblast (HAF) core microtissue generated by gravity-enforced self-assembly in hanging drops, we created a tissue-culture system for studying capillary network formation. We provide comprehensive technical insight into the design and analysis of prototype vascularization in multicell-type-based microtissues. Detailed understanding of generic processes managing capillary formation in human tissue culture may foster advances in the development of clinical tissue implants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CD31(PECAM-1) antibody , Mouse monoclonal, clone WM-59, purified from hybridoma cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Anti-von Willebrand Factor antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution