Merck
All You Ever Wanted to Know About APOL1 and TLFs and Did Not Dare Ask.
Methods in molecular biology (Clifton, N.J.) (2020-03-30)
Joseph Verdi, Charles Schaub, Russell Thomson, Jayne Raper
ABSTRAKT
Interest in trypanosome lytic factors (TLFs) and apolipoprotein L1, the ion channel-forming protein component of TLFs, has increased tenfold since 2010. This is due to the association of African variants of APOL1 with kidney disease such that interest has reached circles beyond parasitology. We have extensive experience purifying and working with these proteins and protein complexes. Herein we describe our detailed purification protocols to aid the new burgeoning field by providing an opportunity for consistency in reagents used across laboratories. We emphasize that it is imperative to maintain APOL1 protein intact (~42 kDa) to analyze the active ion channel-forming component/protein.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Haptoglobin antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder