Merck
Immunotherapeutic approach to cancer with cutaneous DNA vaccination.
Methods in molecular medicine (2001-01-01)
W B Bowne, W G Hawkins, J J Lewis
ABSTRAKT
Innovations for the development of cancer vaccines are emerging from advances in molecular immunology and cancer biology (1). Of these, DNA-based vaccination has become a powerful and potentially versatile method for eliciting an immune response against cancer. One method for DNA immunization involves the delivery of plasmid DNA by particle bombardment. Originally developed for plant hybridization, this approach has proven to be readily transferable to mammalian applications (2-3). Otherwise known as the gene gun, this method allows for the introduction of exogenous "naked" DNA into skin (4) (Fig. 1). Fig. 1. Photograph of the helium-driven gene gun. This is a hand-held device attached to a high pressure helium line and electrical source which operates the trigger. As shown, plastic bullets containing gold particles coated with plasmid DNA are measured and cut to fit within a cartridge. The cartridge, filled with 12 bullets, is easily placed within the barrel of the gun.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%