Merck
Direct conversion of mouse fibroblasts into induced neural stem cells.
Nature protocols (2014-03-22)
Sung Min Kim, Hannah Flaßkamp, Andreas Hermann, Marcos Jesús Araúzo-Bravo, Seung Chan Lee, Sung Ho Lee, Eun Hye Seo, Seung Hyun Lee, Alexander Storch, Hoon Taek Lee, Hans R Schöler, Natalia Tapia, Dong Wook Han
ABSTRAKT
Terminally differentiated cells can be directly converted into different types of somatic cells by using defined factors, thus circumventing the pluripotent state. However, low reprogramming efficiency, along with the absence of proliferation of some somatic cell types, makes it difficult to generate large numbers of cells with this method. Here we describe a protocol to directly convert mouse fibroblasts into self-renewing induced neural stem cells (iNSCs) that can be expanded in vitro, thereby overcoming the limitations associated with low reprogramming efficiency. The four transcription factors required for direct conversion into iNSCs (Sox2, Klf4, Myc (also known as c-Myc) and Pou3f4 (also known as Brn4)) do not generate a pluripotent cell state, and thus the risk for tumor formation after transplantation is reduced. By following the current protocol, iNSCs are observed 4-5 weeks after transduction. Two additional months are required to establish clonal iNSC cell lines that exhibit retroviral transgene silencing and that differentiate into neurons, astrocytes and oligodendrocytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Protamine sulfate salt from salmon, Grade X, amorphous powder
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra