Merck
Detection of Echinococcus granulosus sensu lato infection by using extracts derived from a protoscoleces G1 cell line.
Parasite immunology (2019-09-27)
Andrea Maglioco, Jorge Gentile, Melisa S Barbery Venturi, Oscar Jensen, Claudia Hernández, María Laura Gertiser, Verónica Poggio, Gabriela Canziani, Alicia Graciela Fuchs
ABSTRAKT
Cystic echinococcosis (CE) can be diagnosed by means of several serological approaches, but their results vary among laboratories due to the molecular characteristics of the reference antigens used. Thus, this study aimed to address both the relevance of an EGPE cell line previously obtained from Echinococcus granulosus protoscoleces G1 and the complexity of the immune response by using two different in vitro growth stages as separate sources of parasite antigens. The serum reactivity was investigated by western blotting (WB) in 21 CE patients from an endemic area in a matched case-control design and also in seven experimentally infected sheep and five healthy control sheep. EGPE-antigen-human serum sensitivity by WB was higher than that of hydatid fluid (HF) WB, ELISA and DD5 (P < .05, Chi-square test). EGPE protein extract was immunogenic in mice and hyperimmune plasma reacted with HF proteins, and AgB2 expression was detected by molecular analysis. Proteins of 37 to 60 kDa were recognized by 95.24% of the CE patients' sera but, with poor specificity. Statistically significant differences were found between serum protein extract recognition at 7 and 20 days of cell growth. The EGPE cell line is a laboratory source of antigens for improvement of CE serological diagnosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution