Merck
Quantitative microinjection of mouse oocytes and eggs.
Methods in molecular biology (Clifton, N.J.) (2008-12-17)
Douglas Kline
ABSTRAKT
Quantitative microinjection is used to introduce known quantities of molecules or probes into single cells to examine cellular function. The relatively large mammalian oocyte or egg is easily manipulated and can be injected with impermeant reagents including a variety of signaling molecules and fluorescent probes. Techniques have been developed to inject picoliter quantities of solution into oocytes and eggs with precision and reliability. The methods described here outline the quantitative injection procedures as they are used to inject mouse oocytes and eggs in a culture dish on the stage on an inverted microscope. The techniques are applicable to the oocytes, eggs, and early embryos of most mammalian species. Included are some general instructions on fabrication of transfer pipettes, holding pipettes, beveled injection pipettes, and equipment for quantitative injection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder