Merck
Polarized Human Retinal Pigment Epithelium Exhibits Distinct Surface Proteome on Apical and Basal Plasma Membranes.
Methods in molecular biology (Clifton, N.J.) (2017-12-22)
Vladimir Khristov, Qin Wan, Ruchi Sharma, Mostafa Lotfi, Arvydas Maminishkis, Kapil Bharti
ABSTRAKT
Surface proteins localized on the apical and basal plasma membranes are required for a cell to sense its environment and relay changes in ionic, cytokine, chemokine, and hormone levels to the inside of the cell. In a polarized cell, surface proteins are differentially localized on the apical or the basolateral sides of the cell. The retinal pigment epithelium (RPE) is an example of a polarized cell that performs a variety of functions that are dependent on its polarized state including trafficking of ions, fluid, and metabolites across the RPE monolayer. These functions are absolutely crucial for maintaining the health and integrity of adjacent photoreceptors, the photosensitive cells of the retina. Here we present a series of approaches to identify and validate the polarization state of cultured primary human RPE cells using immunostaining for RPE apical/basolateral markers, polarized cytokine secretion, electrophysiology, fluid transport, phagocytosis, and identification of plasma membrane proteins through cell surface capturing technology. These approaches are currently being used to validate the polarized state and the epithelial phenotype of human induced pluripotent stem (iPS) cell derived RPE cells. This work provides the basis for developing an autologous cell therapy for age-related macular degeneration using patient specific iPS cell derived RPE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
ULTRA Tabletki, Mini, EASYpack Koktajl inhibitorów proteazy, Tablets supplied in foil blister packs.