Merck
Puromycin Labeling Coupled with Proximity Ligation Assays to Define Sites of mRNA Translation in Drosophila Embryos and Human Cells.
Methods in molecular biology (Clifton, N.J.) (2021-10-01)
Ashley Chin, Eric Lécuyer
ABSTRAKT
Genetic mutations, whether they occur within protein-coding or noncoding regions of the genome, can affect various aspects of gene expression by influencing the complex network of intra- and intermolecular interactions that occur between cellular nucleic acids and proteins. One aspect of gene expression control that can be impacted is the intracellular trafficking and translation of mRNA molecules. To study the occurrence and dynamics of translational regulation, researchers have developed approaches such as genome-wide ribosome profiling and artificial reporters that enable single molecule imaging. In this paper, we describe a complementary and optimized approach that combines puromycin labeling with a proximity ligation assay (Puro-PLA) to define sites of translation of specific mRNAs in tissues or cells. This method can be used to study the mechanisms driving the translation of select mRNAs and to access the impact of genetic mutations on local protein synthesis. This approach involves the treatment of cell or tissue specimens with puromycin to label nascently translated peptides, rapid fixation, followed by immunolabeling with appropriate primary and secondary antibodies coupled to PLA oligonucleotide probes, ligation, amplification, and signal detection via fluorescence microscopy. Puro-PLA can be performed at small scale in individual tubes or in chambered slides, or in a high-throughput setup with 96-well plate, for both in situ and in vitro experimentation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture