Merck
Maintenance, Transgene Delivery, and Pluripotency Measurement of Mouse Embryonic Stem Cells.
Methods in molecular biology (Clifton, N.J.) (2015-04-13)
Tetsuya S Tanaka
ABSTRAKT
This chapter describes standard techniques to (1) maintain mouse embryonic stem cell culture, (2) deliver transgenes into mouse embryonic stem cells mediated by electroporation, nucleofection, lipofection, and retro/lentiviruses, and (3) assess the pluripotency of mouse embryonic stem cells. The last part of this chapter presents induction of random cell differentiation followed by the alkaline phosphatase and embryoid body formation assays, immunofluorescence microscopy, and the teratoma formation assay.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Anti-Nestin (C-terminal region) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody