Merck
A ketogenic diet in combination with gemcitabine increases survival in pancreatic cancer KPC mice.
Cancer research communications (2022-11-17)
Natalia E Cortez, Cecilia Rodriguez Lanzi, Brian V Hong, Jihao Xu, Fangyi Wang, Shuai Chen, Jon J Ramsey, Matthew G Pontifex, Michael Müller, David Vauzour, Payam Vahmani, Chang-Il Hwang, Karen Matsukuma, Gerardo G Mackenzie
ABSTRAKT
Pancreatic ductal adenocarcinoma (PDAC) continues to be a major health problem. A ketogenic diet (KD), characterized by a very low carbohydrate and high fat composition, has gained attention for its anti-tumor potential. We evaluated the effect and mechanisms of feeding a strict KD alone or in combination with gemcitabine in the autochthonous LSL-KrasG12D/+; LSL-Trp53 R172H/+; Pdx1-Cre (KPC) mouse model. For this purpose, both male and female pancreatic tumor-bearing KPC mice were allocated to a control diet (CD; %kcal: 70% carb, 14% protein, 16% fat), a KD (%kcal: 14% protein, 1% carb, 85% fat), a CD + gemcitabine (CG), or a KD + gemcitabine (KG) group. Mice fed a KD alone or in combination with gemcitabine showed significantly increased blood β-hydroxybutyrate levels compared to mice fed a CD or CG. KPC mice fed a KG had a significant increase in overall median survival compared to KPC mice fed a CD (increased overall median survival by 42%). Interestingly, when the data was disaggregated by sex, the effect of a KG was significant in female KPC mice (60% increase in median overall survival), but not in male KPC mice (28% increase in median overall survival). Mechanistically, the enhanced survival response to a KD combined with gemcitabine was multifactorial, including inhibition of ERK and AKT pathways, regulation of fatty acid metabolism and the modulation of the gut microbiota. In summary, a KD in combination with gemcitabine appears beneficial as a treatment strategy in PDAC in KPC mice, deserving further clinical evaluation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture