Merck
A genetically encoded far-red fluorescent calcium ion biosensor derived from a biliverdin-binding protein.
Protein science : a publication of the Protein Society (2022-09-30)
Rina Hashizume, Hajime Fujii, Sohum Mehta, Keisuke Ota, Yong Qian, Wenchao Zhu, Mikhail Drobizhev, Yusuke Nasu, Jin Zhang, Haruhiko Bito, Robert E Campbell
ABSTRAKT
Far-red and near-infrared (NIR) genetically encoded calcium ion (Ca2+ ) indicators (GECIs) are powerful tools for in vivo and multiplexed imaging of neural activity and cell signaling. Inspired by a previous report to engineer a far-red fluorescent protein (FP) from a biliverdin (BV)-binding NIR FP, we have developed a far-red fluorescent GECI, designated iBB-GECO1, from a previously reported NIR GECI. iBB-GECO1 exhibits a relatively high molecular brightness, an inverse response to Ca2+ with ΔF/Fmin  = -13, and a near-optimal dissociation constant (Kd ) for Ca2+ of 105 nM. We demonstrate the utility of iBB-GECO1 for four-color multiplexed imaging in MIN6 cells and five-color imaging in HEK293T cells. Like other BV-binding GECIs, iBB-GECO1 did not give robust signals during in vivo imaging of neural activity in mice, but did provide promising results that will guide future engineering efforts. SIGNIFICANCE: Genetically encoded calcium ion (Ca2+ ) indicators (GECIs) compatible with common far-red laser lines (~630-640 nm) on commercial microscopes are of critical importance for their widespread application to deep-tissue multiplexed imaging of neural activity. In this study, we engineered a far-red excitable fluorescent GECI, designated iBB-GECO1, that exhibits a range of preferable specifications such as high brightness, large fluorescence response to Ca2+ , and compatibility with multiplexed imaging in mammalian cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline