Merck
Necroptotic virotherapy of oncolytic alphavirus M1 cooperated with Doxorubicin displays promising therapeutic efficacy in TNBC.
Oncogene (2021-06-23)
Jiayu Zhang, Ying Liu, Jingyi Tan, Yanming Zhang, Chun-Wa Wong, Ziqing Lin, Xincheng Liu, Max Sander, Xiaozhi Yang, Lebin Liang, Deli Song, Jia Dan, Yuwei Zhou, Jing Cai, Yuan Lin, Jiankai Liang, Jun Hu, Guangmei Yan, Wenbo Zhu
ABSTRAKT
Triple-negative breast cancer (TNBC) is the most aggressive molecular subtype among breast tumors and remains a challenge even for the most current therapeutic regimes. Here, we demonstrate that oncolytic alphavirus M1 effectively kills both TNBC and non-TNBC. ER-stress and apoptosis pathways are responsible for the cell death in non-TNBC as reported in other cancer types, yet the cell death in TNBC does not depend on these pathways. Transcriptomic analysis reveals that the M1 virus activates necroptosis in TNBC, which can be pharmacologically blocked by necroptosis inhibitors. By screening a library of clinically available compounds commonly used for breast cancer treatment, we find that Doxorubicin enhances the oncolytic effect of the M1 virus by up to 100-fold specifically in TNBC in vitro, and significantly stalls the tumor growth of TNBC in vivo, through promoting intratumoral virus replication and further triggering apoptosis in addition to necroptosis. These findings reveal a novel antitumor mechanism and a new combination regimen of the M1 oncolytic virus in TNBC, and highlight a need to bridge molecular diagnosis with virotherapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HS-1371 trifluoroacetate salt, ≥98% (HPLC)