Merck
In vivo assay of cellular proliferation.
Methods in molecular biology (Clifton, N.J.) (2001-05-19)
J Curran
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade