Merck
Cell detachment rates and confluence of fibroblast and osteoblast cell culture using different washing solutions.
Brazilian journal of biology = Revista brasleira de biologia (2023-01-27)
H S Budi, M C Setyawati, S Anitasari, Y-K Shen, I Pebriani, D E Ramadan
ABSTRAKT
The advancements in the cell culture studies have led to the development of regenerative medicine concept. The aim of this study is to compare the effectiveness of some washing solutions, including phosphate buffered saline (PBS), sodium chloride (NaCl), and ringer's lactate (RL) on the rate of detachment and confluency in fibroblast and osteoblast cell culture. Baby Hamster Kidney 21 clone 13 (BHK21/C13) fibroblast cells and 7F2 osteoblast were cultured on T25 flasks for 3-4 days. Three treatment groups were classified on the basis of different washing solutions used in the moment before trypsinization: PBS, 0.9% NaCl, and RL. Each group was measured for the detachment rate and cell confluence. The measurement was done in 2 passage numbers. The use of PBS, NaCl, and RL washing solution showed that detachment time was less than 5 minutes for the fibroblasts and 3 minutes for the osteoblasts. There was a significant difference in the rate of fibroblast cell detachment (p=0.006) and osteoblast (p=0.016). The capability of fibroblasts and osteoblasts to achieve a confluence of 106 cells/well on the first and second measurements was almost the same between the washing solution groups. The use of physiological 0.9% NaCl solution as a washing solution in fibroblast and osteoblast cell culture has almost the same effectiveness as PBS to help accelerate cell detachment in less than 5 minutes without influencing the capability of cells to proliferate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red