Merck
Potassium Iodide Nanoparticles Enhance Radiotherapy against Breast Cancer by Exploiting the Sodium-Iodide Symporter.
ACS nano (2021-10-26)
Benjamin L Cline, Wen Jiang, Chaebin Lee, Zhengwei Cao, Xueyuan Yang, Shuyue Zhan, Harrison Chong, Tao Zhang, Zhaoguo Han, Xuedan Wu, Li Yao, Hui Wang, Weizhong Zhang, Zibo Li, Jin Xie
ABSTRAKT
Iodine has shown promise in enhancing radiotherapy. However, conventional iodine compounds show fast clearance and low retention inside cancer cells, limiting their application as a radiosensitizer. Herein, we synthesize poly(maleic anhydride-alt-1-octadecene) coated KI nanoparticles (PMAO-KI NPs) and evaluate their potential for enhancing radiotherapy. Owing to the polymer coating, the KI core of PMAO-KI NPs is not instantly dissolved in aqueous solutions but slowly degraded, allowing for controlled release of iodide (I-). I- is transported into cells via the sodium iodide symporter (NIS), which is upregulated in breast cancer cells. Our results show that PMAO-KI NPs can enhance radiation-induced production of reactive oxygen species such as hydroxyl radicals. When tested in vitro with MCF-7 cells, PMAO-KI NPs promote radiation-induced DNA double-strand breaks and lipid peroxidation, causing a drop in cancer cell viability and reproductivity. When tested in MCF-7 bearing mice, PMAO-KI NPs show significant radiosensitizing effects, leading to complete tumor eradication in 80% of the treated animals without inducing additional toxicity. Overall, our strategy exploits electrolyte nanoparticles to deliver iodide into cancer cells through NIS, thus promoting radiotherapy against breast cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo H2A.X (Ser139), koniugat Alexa 647, from rabbit, ALEXA FLUOR 647