Merck
Small-molecule activation of OGG1 increases oxidative DNA damage repair by gaining a new function.
Science (New York, N.Y.) (2022-06-24)
Maurice Michel, Carlos Benítez-Buelga, Patricia A Calvo, Bishoy M F Hanna, Oliver Mortusewicz, Geoffrey Masuyer, Jonathan Davies, Olov Wallner, Kumar Sanjiv, Julian J Albers, Sergio Castañeda-Zegarra, Ann-Sofie Jemth, Torkild Visnes, Ana Sastre-Perona, Akhilesh N Danda, Evert J Homan, Karthick Marimuthu, Zhao Zhenjun, Celestine N Chi, Antonio Sarno, Elisée Wiita, Catharina von Nicolai, Anna J Komor, Varshni Rajagopal, Sarah Müller, Emily C Hank, Marek Varga, Emma R Scaletti, Monica Pandey, Stella Karsten, Hanne Haslene-Hox, Simon Loevenich, Petra Marttila, Azita Rasti, Kirill Mamonov, Florian Ortis, Fritz Schömberg, Olga Loseva, Josephine Stewart, Nicholas D'Arcy-Evans, Tobias Koolmeister, Martin Henriksson, Dana Michel, Ana de Ory, Lucia Acero, Oriol Calvete, Martin Scobie, Christian Hertweck, Ivan Vilotijevic, Christina Kalderén, Ana Osorio, Rosario Perona, Alexandra Stolz, Pål Stenmark, Ulrika Warpman Berglund, Miguel de Vega, Thomas Helleday
ABSTRAKT
Oxidative DNA damage is recognized by 8-oxoguanine (8-oxoG) DNA glycosylase 1 (OGG1), which excises 8-oxoG, leaving a substrate for apurinic endonuclease 1 (APE1) and initiating repair. Here, we describe a small molecule (TH10785) that interacts with the phenylalanine-319 and glycine-42 amino acids of OGG1, increases the enzyme activity 10-fold, and generates a previously undescribed β,δ-lyase enzymatic function. TH10785 controls the catalytic activity mediated by a nitrogen base within its molecular structure. In cells, TH10785 increases OGG1 recruitment to and repair of oxidative DNA damage. This alters the repair process, which no longer requires APE1 but instead is dependent on polynucleotide kinase phosphatase (PNKP1) activity. The increased repair of oxidative DNA lesions with a small molecule may have therapeutic applications in various diseases and aging.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
TH10785, ≥98% (HPLC)