Merck
Loss of GLTSCR1 causes congenital heart defects by regulating NPPA transcription.
Angiogenesis (2023-02-07)
Fengyan Han, Beibei Yang, Yan Chen, Lu Liu, Xiaoqing Cheng, Jiaqi Huang, Ke Zhou, Dandan Zhang, Enping Xu, Maode Lai, Bingjian Lv, Hongqiang Cheng, Honghe Zhang
ABSTRAKT
Precise and specific spatiotemporal domains of gene expression regulation are critical for embryonic development. Recent studies have identified GLTSCR1 as a gene transcriptional elongation regulator in cancer research. However, the function of GLTSCR1, especially in embryonic development, remains poorly understood. Here, we found that GLTSCR1 was essential for cardiac development because Gltscr1 knockout (Gltscr1-/-) led to embryonic lethality in mice with severe congenital heart defects (CHDs). Ventricular septal defect and double outflow right ventricular were also observed in neural crest cells with conditional deletion of Gltscr1, which were associated with neonatal lethality in mice. Mechanistically, GLTSCR1 deletion promoted NPPA expression by coordinating the CHD risk G allele of rs56153133 in the NPPA enhancer and releasing the transcription factor ZNF740-binding site on the NPPA promoter. These findings demonstrated that GLTSCR1 acts as a candidate CHD-related gene.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody