Merck
  • LncRNA KCNQ1OT1 contributes to hydrogen peroxide-induced apoptosis, inflammation, and oxidative stress of cardiomyocytes via miR-130a-3p/ZNF791 axis.
LncRNA KCNQ1OT1 contributes to hydrogen peroxide-induced apoptosis, inflammation, and oxidative stress of cardiomyocytes via miR-130a-3p/ZNF791 axis.
Cell biology international (2022-08-23)
Hong Xin, Chengliang Li, Tianzhi Cai, Jinlong Cao, Meixue Wang
ABSTRAKT
It has been reported that long noncoding RNA (lncRNA) KCNQ1 opposite strand/antisense transcript 1 (KCNQ1OT1) played an important role in myocardial infarction (MI). However, the regulatory network behind KCNQ1OT1 in MI is largely unknown. Quantitative real time polymerase chain reaction (qRT-PCR) was applied to detect the enrichment of KCNQ1OT1, microRNA-130a-3p (miR-130a-3p) and zinc finger 791 (ZNF791). The viability and apoptosis of AC16 cells were measured by (4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay and flow cytometry. Enzyme-linked immunosorbent assay (ELISA) was conducted to assess the inflammation and oxidative stress status of AC16 cells. The targeted relationship between miR-130a-3p and KCNQ1OT1 or ZNF791 was predicted by StarBase bioinformatic database, and dual-luciferase reporter assay and RNA immunoprecipitation (RIP) assay were carried out to verify these predictions. Hydrogen peroxide (H2 O2 ) stimulation caused a significant upregulation in the expression of KCNQ1OT1, while the level of miR-130a-3p showed an opposite phenomenon. KCNQ1OT1 was a crucial downstream component in H2 O2 -mediated toxic effects, and KCNQ1OT1 accelerated H2 O2 -induced toxic effects in AC16 cells. KCNQ1OT1 could sponge miR-130a-3p and down-regulate its expression. MiR-130a-3p exerted opposite effects to KCNQ1OT1, and the depletion of miR-130a-3p attenuated the protective effects of KCNQ1OT1 intervention on AC16 cells exposed to H2 O2 . MiR-130a-3p could bind to ZNF791, and ZNF791 served as the target of miR-130a-3p to promote H2 O2 -induced injury of AC16 cells. ZNF791 was modulated by KCNQ1OT1/miR-130a-3p signaling in H2 O2 -treated AC16 cells. In all, lncRNA KCNQ1OT1 deteriorated H2 O2 -mediated injury in cardiomyocytes through upregulating ZNF791 via serving as a molecular sponge for miR-130a-3p.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-ZNF791 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution