Merck
Establishment and Use of Mouse Haploid ES Cells.
Current protocols in mouse biology (2015-06-13)
Martin Leeb, Anthony C F Perry, Anton Wutz
ABSTRAKT
Haploid genetics has facilitated new insights into mammalian pathways and disease mechanisms. Most animal cells are diploid, and mammalian haploid cell cultures have remained elusive for a long time. Recent methodological progress has enabled the routine derivation of haploid stem cell lines from mammalian haploid embryos. Here we provide detailed protocols for the establishment, culture, and manipulation of parthenogenetic and androgenetic haploid embryonic stem cells from mouse embryos.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)