Merck
  • Discovery of a 1-Methyl-3,4-dihydronaphthalene-Based Sphingosine-1-Phosphate (S1P) Receptor Agonist Ceralifimod (ONO-4641). A S1P1 and S1P5 Selective Agonist for the Treatment of Autoimmune Diseases.
Discovery of a 1-Methyl-3,4-dihydronaphthalene-Based Sphingosine-1-Phosphate (S1P) Receptor Agonist Ceralifimod (ONO-4641). A S1P1 and S1P5 Selective Agonist for the Treatment of Autoimmune Diseases.
Journal of medicinal chemistry (2017-11-10)
Haruto Kurata, Kensuke Kusumi, Kazuhiro Otsuki, Ryo Suzuki, Masakuni Kurono, Takaki Komiya, Hiroshi Hagiya, Hirotaka Mizuno, Hiroki Shioya, Takeji Ono, Yuka Takada, Tatsuo Maeda, Norikazu Matsunaga, Tetsu Kondo, Sachiko Tominaga, Ken-Ici Nunoya, Hidekazu Kiyoshi, Masaharu Komeno, Shinji Nakade, Hiromu Habashita
ABSTRAKT
The discovery of 1-({6-[(2-methoxy-4-propylbenzyl)oxy]-1-methyl-3,4-dihydronaphthalen-2-yl}methyl)azetidine-3-carboxylic acid 13n (ceralifimod, ONO-4641), a sphingosine-1-phosphate (S1P) receptor agonist selective for S1P1 and S1P5, is described. While it has been revealed that the modulation of the S1P1 receptor is an effective way to treat autoimmune diseases such as relapsing-remitting multiple sclerosis (RRMS), it was also reported that activation of the S1P3 receptor is implicated in some undesirable effects. We carried out a structure-activity relationship (SAR) study of hit compound 6 with an amino acid moiety in the hydrophilic head region. Following identification of a lead compound with a dihydronaphthalene central core by inducing conformational constraint, optimization of the lipophilic tail region led to the discovery of 13n as a clinical candidate that exhibited >30 000-fold selectivity for S1P1 over S1P3 and was potent in a peripheral lymphocyte lowering (PLL) test in mice (ED50 = 0.029 mg/kg, 24 h after oral dosing).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ceralifimod hydrochloride, ≥95% (HPLC)