Merck
Human WDR5 promotes breast cancer growth and metastasis via KMT2-independent translation regulation.
eLife (2022-09-01)
Wesley L Cai, Jocelyn Fang-Yi Chen, Huacui Chen, Emily Wingrove, Sarah J Kurley, Lok Hei Chan, Meiling Zhang, Anna Arnal-Estape, Minghui Zhao, Amer Balabaki, Wenxue Li, Xufen Yu, Ethan D Krop, Yali Dou, Yansheng Liu, Jian Jin, Thomas F Westbrook, Don X Nguyen, Qin Yan
ABSTRAKT
Metastatic breast cancer remains a major cause of cancer-related deaths in women, and there are few effective therapies against this advanced disease. Emerging evidence suggests that key steps of tumor progression and metastasis are controlled by reversible epigenetic mechanisms. Using an in vivo genetic screen, we identified WDR5 as an actionable epigenetic regulator that is required for metastatic progression in models of triple-negative breast cancer. We found that knockdown of WDR5 in breast cancer cells independently impaired their tumorigenic as well as metastatic capabilities. Mechanistically, WDR5 promotes cell growth by increasing ribosomal gene expression and translation efficiency in a KMT2-independent manner. Consistently, pharmacological inhibition or degradation of WDR5 impedes cellular translation rate and the clonogenic ability of breast cancer cells. Furthermore, a combination of WDR5 targeting with mTOR inhibitors leads to potent suppression of translation and proliferation of breast cancer cells. These results reveal novel therapeutic strategies to treat metastatic breast cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Roche
cOmplete, Mini koktajl inhibitorów proteaz, Tablets provided in a glass vial
Sigma-Aldrich
MS67N, ≥98% (HPLC)
Sigma-Aldrich
MS67, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)