Merck
Structure of dimeric lipoprotein lipase reveals a pore adjacent to the active site.
Nature Communications (2023)
Gunn, et al.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Lipoprotein Lipase/LPL Antibody, clone 4-1a, clone 4-1a, from mouse