- Integrative Single-Cell Transcriptomics and Epigenomics Mapping of the Fetal Retina Developmental Dynamics.
Integrative Single-Cell Transcriptomics and Epigenomics Mapping of the Fetal Retina Developmental Dynamics.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2023)
Li, et al.
PMID37017569