Merck
Aberrant Wnt Signaling Induces Comedo-Like Changes in the Murine Upper Hair Follicle.
The Journal of Investigative Dermatology (2022)
Shang, et al.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)