Merck
Mouse Models for Atopic Dermatitis.
Current protocols (2023)
Sakamoto, et al.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein