Merck
De novo missense variants in RRAGC lead to a fatal mTORopathy of early childhood.
Genetics in medicine : official journal of the American College of Medical Genetics (2023-04-15)
Margot R F Reijnders, Annette Seibt, Melanie Brugger, Ideke J C Lamers, Torsten Ott, Oliver Klaas, Judit Horváth, Ailsa M S Rose, Isabel M Craghill, Theresa Brunet, Elisabeth Graf, Katharina Mayerhanser, Debby Hellebrekers, David Pauck, Eva Neuen-Jacob, Richard J T Rodenburg, Dagmar Wieczorek, Dirk Klee, Ertan Mayatepek, Gertjan Driessen, Robert Bindermann, Luisa Averdunk, Klaus Lohmeier, Margje Sinnema, Alexander P A Stegmann, Ronald Roepman, James A Poulter, Felix Distelmaier
ABSTRAKT
Mechanistic target of rapamycin (mTOR) complex 1 (mTORC1) regulates cell growth in response to nutritional status. Central to the mTORC1 function is the Rag-GTPase heterodimer. One component of the Rag heterodimer is RagC (Ras-related GTP-binding protein C), which is encoded by the RRAGC gene. Genetic testing via trio exome sequencing was applied to identify the underlying disease cause in 3 infants with dilated cardiomyopathy, hepatopathy, and brain abnormalities, including pachygyria, polymicrogyria, and septo-optic dysplasia. Studies in patient-derived skin fibroblasts and in a HEK293 cell model were performed to investigate the cellular consequences. We identified 3 de novo missense variants in RRAGC (NM_022157.4: c.269C>A, p.(Thr90Asn), c.353C>T, p.(Pro118Leu), and c.343T>C, p.(Trp115Arg)), which were previously reported as occurring somatically in follicular lymphoma. Studies of patient-derived fibroblasts carrying the p.(Thr90Asn) variant revealed increased cell size, as well as dysregulation of mTOR-related p70S6K (ribosomal protein S6 kinase 1) and transcription factor EB signaling. Moreover, subcellular localization of mTOR was decoupled from metabolic state. We confirmed the key findings for all RRAGC variants described in this study in a HEK293 cell model. The above results are in line with a constitutive overactivation of the mTORC1 pathway. Our study establishes de novo missense variants in RRAGC as cause of an early-onset mTORopathy with unfavorable prognosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-Lamin A/C antibody produced in mouse, clone 4C11, purified from hybridoma cell culture