Merck
Repurposing a tricyclic antidepressant in tumor and metabolism disease treatment through fatty acid uptake inhibition.
The Journal of experimental medicine (2022-12-16)
Qiaoyun Chu, Jing An, Ping Liu, Yihan Song, Xuewei Zhai, Ronghui Yang, Jing Niu, Chuanzhen Yang, Binghui Li
ABSTRAKT
Fatty acid uptake is essential for cell physiological function, but detailed mechanisms remain unclear. Here, we generated an acetyl-CoA carboxylases (ACC1/2) double-knockout cell line, which lacked fatty acid biosynthesis and survived on serum fatty acids and was used to screen for fatty acid uptake inhibitors. We identified a Food and Drug Administration-approved tricyclic antidepressant, nortriptyline, that potently blocked fatty acid uptake both in vitro and in vivo. We also characterized underlying mechanisms whereby nortriptyline provoked lysosomes to release protons and induce cell acidification to suppress macropinocytosis, which accounted for fatty acid endocytosis. Furthermore, nortriptyline alone or in combination with ND-646, a selective ACC1/2 inhibitor, significantly repressed tumor growth, lipogenesis, and hepatic steatosis in mice. Therefore, we show that cells actively take up fatty acids through macropinocytosis, and we provide a potential strategy suppressing tumor growth, lipogenesis, and hepatic steatosis through controlling the cellular level of fatty acids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture