Merck
Genome-wide mapping of cancer dependency genes and genetic modifiers of chemotherapy in high-risk hepatoblastoma.
Nature communications (2023-07-07)
Jie Fang, Shivendra Singh, Changde Cheng, Sivaraman Natarajan, Heather Sheppard, Ahmed Abu-Zaid, Adam D Durbin, Ha Won Lee, Qiong Wu, Jacob Steele, Jon P Connelly, Hongjian Jin, Wenan Chen, Yiping Fan, Shondra M Pruett-Miller, Jerold E Rehg, Selene C Koo, Teresa Santiago, Joseph Emmons, Stefano Cairo, Ruoning Wang, Evan S Glazer, Andrew J Murphy, Taosheng Chen, Andrew M Davidoff, Carolina Armengol, John Easton, Xiang Chen, Jun Yang
ABSTRAKT
A lack of relevant genetic models and cell lines hampers our understanding of hepatoblastoma pathogenesis and the development of new therapies for this neoplasm. Here, we report an improved MYC-driven hepatoblastoma-like murine model that recapitulates the pathological features of embryonal type of hepatoblastoma, with transcriptomics resembling the high-risk gene signatures of the human disease. Single-cell RNA-sequencing and spatial transcriptomics identify distinct subpopulations of hepatoblastoma cells. After deriving cell lines from the mouse model, we map cancer dependency genes using CRISPR-Cas9 screening and identify druggable targets shared with human hepatoblastoma (e.g., CDK7, CDK9, PRMT1, PRMT5). Our screen also reveals oncogenes and tumor suppressor genes in hepatoblastoma that engage multiple, druggable cancer signaling pathways. Chemotherapy is critical for human hepatoblastoma treatment. A genetic mapping of doxorubicin response by CRISPR-Cas9 screening identifies modifiers whose loss-of-function synergizes with (e.g., PRKDC) or antagonizes (e.g., apoptosis genes) the effect of chemotherapy. The combination of PRKDC inhibition and doxorubicin-based chemotherapy greatly enhances therapeutic efficacy. These studies provide a set of resources including disease models suitable for identifying and validating potential therapeutic targets in human high-risk hepatoblastoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
JumpStart Taq DNA Polymerase, with MgCl2
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid