Merck
Sex differences in proliferation and attrition of pubertally born cells in the rat posterior dorsal medial amygdala.
Developmental cognitive neuroscience (2022-08-08)
Margaret A Mohr, Nancy Staffend Michael, Lydia L DonCarlos, Cheryl L Sisk
ABSTRAKT
The rodent posterodorsal medial amygdala (MePD) evaluates and assigns valence to social sensory stimuli. The perception of social stimuli evolves during puberty, when the focus of social interactions shifts from kin to peers. Using the cell birthdate marker bromo-deoxyuridine (BrdU), we previously discovered that more pubertally born cells are added to the rat MePD in males than females. Here we addressed several questions that remained unanswered by our previous work. First, to determine whether there are sex differences in cell proliferation within the MePD, we examined BrdU-immunoreactive (-ir) cells at 2 and 4 h following BrdU administration on postnatal day 30 (P30). The density of BrdU-ir cells was greater in males than in females, indicating greater proliferation in males. Proliferation was substantiated by double-label immunohistochemistry showing that MePD BrdU-ir cells colocalize proliferating cell nuclear antigen, but not the cell death marker Caspase3. We next studied longer time points (2-21 days) following BrdU administration on P30 and found that the rate of cell attrition is higher in males. Finally, triple-label immunohistochemistry of P30-born MePD cells revealed that some of these cells differentiate into neurons or astrocytes within three weeks of cell birth, with no discernable sex differences. The demonstration of pubertal neuro- and glio-genesis in the MePD of male and female rats adds a new dimension to developmental plasticity of the MePD that may contribute to pubertal changes in the perception of social stimuli in both sexes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse