- Orphan quality control shapes network dynamics and gene expression.
Orphan quality control shapes network dynamics and gene expression.
Cell (2023-09-16)
Kevin G Mark, SriDurgaDevi Kolla, Jacob D Aguirre, Danielle M Garshott, Stefan Schmitt, Diane L Haakonsen, Christina Xu, Lukas Kater, Georg Kempf, Brenda Martínez-González, David Akopian, Stephanie K See, Nicolas H Thomä, Michael Rapé
PMID37714136
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Roche
cOmplete™™ Mini, Koktajl inhibitorów proteaz bez EDTA, Tablets provided in EASYpacks