Merck
Topographical distribution and morphology of SP-IR axons in the antrum, pylorus, and duodenum of mice.
Autonomic neuroscience : basic & clinical (2023-02-23)
Anas Mistareehi, Kohlton T Bendowski, Ariege Bizanti, Jazune Madas, Yuanyuan Zhang, Andrew M Kwiat, Duyen Nguyen, Nicole Kogut, Jichao Ma, Jin Chen, Zixi Jack Cheng
ABSTRAKT
Substance-P (SP) is a commonly used marker of nociceptive afferent axons, and it plays an important role in a variety of physiological functions including the regulation of motility, gut secretion, and vascular flow. Previously, we found that SP-immunoreactive (SP-IR) axons densely innervated the pyloric antrum of the flat-mount of the mouse whole stomach muscular layer. However, the regional distribution and morphology of SP-IR axons in the submucosa and mucosa were not well documented. In this study, the mouse antrum-pylorus-duodenum (APD) were transversely and longitudinally sectioned. A Zeiss M2 imager was used to scan the serial sections of each APD (each section montage consisted of 50-100 all-in-focus maximal projection images). To determine the detailed structures of SP-IR axons and terminals, we used the confocal microscope to scan the regions of interest. We found that 1) SP-IR axons innervated the muscular, submucosal, and mucosal layers. 2) In the muscular layer, SP-IR varicose axons densely innervated the muscles and formed varicose terminals which encircled myenteric neurons. 3) In the submucosa, SP-IR axons innervated blood vessels and submucosal ganglia and formed a network in Brunner's glands. 4) In the mucosa, SP-IR axons innervated the muscularis mucosae. Some SP-IR axons entered the lamina propria. 5) The muscular layer of the antrum and duodenum showed a higher SP-IR axon density than the pyloric sphincter. 6) SP-IR axons were from extrinsic and intrinsic origins. This work provided a comprehensive view of the distribution and morphology of SP-IR axons in the APD at single cell/axon/varicosity scale. This data will be used to create a 3D scaffold of the SP-IR axon innervation of the APD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody, from goat, purified by affinity chromatography
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from sheep