Merck
Efficient Large DNA Fragment Knock-in by Long dsDNA with 3'-Overhangs Mediated CRISPR Knock-in (LOCK) in Mammalian Cells.
Bio-protocol (2023-10-30)
Wenjie Han, Haojun Liang, Jianqiang Bao
ABSTRAKT
An efficient and precise genome-editing approach is in high demand in any molecular biology or cell biology laboratory worldwide. However, despite a recent rapid progress in the toolbox tailored for precise genome-editing, including the base editors and prime editors, there is still a need for a cost-effective knock-in (KI) approach amenable for long donor DNA cargos with high efficiency. By harnessing the high-efficient double-strand break (DSB) repair pathway of microhomology-mediated end joining, we previously showed that a specially designed 3'-overhang double-strand DNA (odsDNA) donor harboring 50-nt homology arm (HA) allows high-efficient exogenous DNA KI when combined with CRISPR-Cas9 technology. The lengths of the 3'-overhangs of odsDNA donors could be manipulated by the five consecutive phosphorothioate (PT) modifications. In this protocol, we detail the stepwise procedures to conduct the LOCK (Long dsDNA with 3'-Overhangs mediated CRISPR Knock-in) method for gene-sized (~1-3 kb) KI in mammalian cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rosetta 2(DE3) Competent Cells - Novagen, Novagen′s Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.