Merck
SCFβTrCP-mediated degradation of SHARP1 in triple-negative breast cancer.
Cell death & disease (2023-11-08)
Juliana Haydeé Enriqué Steinberg, Fabiana Alejandra Rossi, Roberto Magliozzi, Laurensia Yuniati, Matteo Santucci, Mario Rossi, Daniele Guardavaccaro, Angela Lauriola
ABSTRAKT
Triple-negative breast cancer (TNBC) is a subtype of breast cancer associated with metastasis, high recurrence rate, and poor survival. The basic helix-loop-helix transcription factor SHARP1 (Split and Hairy-related Protein 1) has been identified as a suppressor of the metastatic behavior of TNBC. SHARP1 blocks the invasive phenotype of TNBC by inhibiting hypoxia-inducible factors and its loss correlates with poor survival of breast cancer patients. Here, we show that SHARP1 is an unstable protein that is targeted for proteasomal degradation by the E3 ubiquitin ligase complex SCFβTrCP. SHARP1 recruits βTrCP via a phosphodegron encompassing Ser240 and Glu245 which are required for SHARP1 ubiquitylation and degradation. Furthermore, mice injected with TNBC cells expressing the non-degradable SHARP1(S240A/E245A) mutant display reduced tumor growth and increased tumor-free survival. Our study suggests that targeting the βTrCP-dependent degradation of SHARP1 represents a therapeutic strategy in TNBC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)