Merck
A CRISPR-drug perturbational map for identifying compounds to combine with commonly used chemotherapeutics.
Nature communications (2023-11-14)
Hyeong-Min Lee, William C Wright, Min Pan, Jonathan Low, Duane Currier, Jie Fang, Shivendra Singh, Stephanie Nance, Ian Delahunty, Yuna Kim, Richard H Chapple, Yinwen Zhang, Xueying Liu, Jacob A Steele, Jun Qi, Shondra M Pruett-Miller, John Easton, Taosheng Chen, Jun Yang, Adam D Durbin, Paul Geeleher
ABSTRAKT
Combination chemotherapy is crucial for successfully treating cancer. However, the enormous number of possible drug combinations means discovering safe and effective combinations remains a significant challenge. To improve this process, we conduct large-scale targeted CRISPR knockout screens in drug-treated cells, creating a genetic map of druggable genes that sensitize cells to commonly used chemotherapeutics. We prioritize neuroblastoma, the most common extracranial pediatric solid tumor, where ~50% of high-risk patients do not survive. Our screen examines all druggable gene knockouts in 18 cell lines (10 neuroblastoma, 8 others) treated with 8 widely used drugs, resulting in 94,320 unique combination-cell line perturbations, which is comparable to the largest existing drug combination screens. Using dense drug-drug rescreening, we find that the top CRISPR-nominated drug combinations are more synergistic than standard-of-care combinations, suggesting existing combinations could be improved. As proof of principle, we discover that inhibition of PRKDC, a component of the non-homologous end-joining pathway, sensitizes high-risk neuroblastoma cells to the standard-of-care drug doxorubicin in vitro and in vivo using patient-derived xenograft (PDX) models. Our findings provide a valuable resource and demonstrate the feasibility of using targeted CRISPR knockout to discover combinations with common chemotherapeutics, a methodology with application across all cancers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
AC16 Human Cardiomyocyte Cell Line, AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes.