Merck
Bidirectional movement of tunicamycin in Arabidopsis thaliana.
The New phytologist (2023-11-02)
Anh H Ngo, Yu-Ching Wu, Yuki Nakamura
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tunicamycin, Streptomyces lysosuperficus