Merck
  • Generation and characterization of human induced pluripotent stem cells (iPSCs) from three patients with age-related macular degeneration carrying rare variants in the CFH gene.
Generation and characterization of human induced pluripotent stem cells (iPSCs) from three patients with age-related macular degeneration carrying rare variants in the CFH gene.
Stem cell research (2022-01-23)
Louet Koolen, Giuliana Gagliardi, Sofie C A Ten Brink, Anita de Breuk, Thomas J Heesterbeek, Carel B Hoyng, Silvia Albert, Anneke I den Hollander
ABSTRAKT
Age-related macular degeneration (AMD) is a major cause of vision loss among the elderly in the Western world. AMD is multifactorial eye disease with a strong genetic contribution. Here, we report the generation and characterization of induced pluripotent stem cells (iPSCs) derived from peripheral blood mononuclear cells of three patients with AMD carrying rare variants in the complement factor H (CFH) gene. These cell lines were generated for cellular studies investigating the disease mechanisms and developing therapeutic interventions for AMD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse