Merck
Design and synthesis of 4-Aminoquinoline-isoindoline-dione-isoniazid triads as potential anti-mycobacterials.
Bioorganic & medicinal chemistry letters (2020-09-28)
Anu Rani, Matt D Johansen, Françoise Roquet-Banères, Laurent Kremer, Paul Awolade, Oluwakemi Ebenezer, Parvesh Singh, Sumanjit, Vipan Kumar
ABSTRAKT
A series of 4-aminoquinoline-isoindoline-dione-isoniazid triads were synthesized and assessed for their anti-mycobacterial activities and cytotoxicity. Most of the synthesized compounds exhibited promising activities against the mc26230 strain of M. tuberculosis with MIC in the range of 5.1-11.9 µM and were non-cytotoxic against Vero cells. The conjugates lacking either isoniazid or quinoline core in their structural framework failed to inhibit the growth of M. tuberculosis; thus, further strengthening the proposed design of triads in the present study.