Merck
Cerebrospinal fluid calcium homeostasis: evidence for a plasma membrane Ca2+-pump in mammalian choroid plexus.
Brain research (1989-06-12)
J L Borke, A J Caride, T L Yaksh, J T Penniston, R Kumar
ABSTRAKT
A major unanswered question in central nervous system physiology concerns the mechanism by which cerebrospinal fluid (CSF) Ca2+ homeostasis is maintained in the face of hypo- or hypercalcemia. To address this question, we sought and found a protein of Mr approximately 140,000 in choroid plexus plasma membranes that forms a phosphorylated intermediate with characteristics of a plasma membrane Ca2+-pump. A choroid plexus plasma membrane protein of this molecular weight also bound to a monoclonal antibody prepared against the human erythrocyte plasma membrane Ca2+-Mg2+ ATPase Ca2+-pump. When this monoclonal antibody was used for immunohistochemical localization, the plasma membrane Ca2+-pump was found primarily in the CSF-facing membranes of choroid plexus cells from rats, cats, and man. The localization of a plasma membrane Ca2+-pump in the CSF-facing membranes of the choroid plexus suggests that the choroid plexus, by mechanisms including this pump, may regulate CSF Ca2+ concentrations.