Merck
Distinct forebrain regions define a dichotomous astrocytic profile in multiple system atrophy.
Acta neuropathologica communications (2024-01-04)
Y Schneider, C Gauer, M Andert, A Hoffmann, M J Riemenschneider, W Krebs, N Chalmers, C Lötzsch, U J Naumann, W Xiang, V Rothhammer, R Beckervordersandforth, J C M Schlachetzki, J Winkler
ABSTRAKT
The growing recognition of a dichotomous role of astrocytes in neurodegenerative processes has heightened the need for unraveling distinct astrocytic subtypes in neurological disorders. In multiple system atrophy (MSA), a rare, rapidly progressing atypical Parkinsonian disease characterized by increased astrocyte reactivity. However the specific contribution of astrocyte subtypes to neuropathology remains elusive. Hence, we first set out to profile glial fibrillary acidic protein levels in astrocytes across the human post mortem motor cortex, putamen, and substantia nigra of MSA patients and observed an overall profound astrocytic response. Matching the post mortem human findings, a similar astrocytic phenotype was present in a transgenic MSA mouse model. Notably, MSA mice exhibited a decreased expression of the glutamate transporter 1 and glutamate aspartate transporter in the basal ganglia, but not the motor cortex. We developed an optimized astrocyte isolation protocol based on magnetic-activated cell sorting via ATPase Na+/K+ transporting subunit beta 2 and profiled the transcriptomic landscape of striatal and cortical astrocytes in transgenic MSA mice. The gene expression profile of astrocytes in the motor cortex displayed an anti-inflammatory signature with increased oligodendroglial and pro-myelinogenic expression pattern. In contrast, striatal astrocytes were defined by elevated pro-inflammatory transcripts accompanied by dysregulated genes involved in homeostatic functions for lipid and calcium metabolism. These findings provide new insights into a region-dependent, dichotomous astrocytic response-potentially beneficial in the cortex and harmful in the striatum-in MSA suggesting a differential role of astrocytes in MSA-related neurodegenerative processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Water Channel Aquaporin 4 antibody produced in rabbit, lyophilized powder, affinity isolated antibody
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Glutamate Transporter Antibody, Glial, serum, Chemicon®
Sigma-Aldrich
Anti-Growth Associated Protein 43 Antibody, clone 9-1E12, clone 9-1E12, Chemicon®, from mouse