Merck
In vitro efficacy of Coriandrum sativum, Lippia sidoides and Copaifera reticulata against Leishmania chagasi.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria (2012-10-17)
Fernanda Cristina Macedo Rondon, Claudia Maria Leal Bevilaqua, Marina Parissi Accioly, Selene Maia de Morais, Heitor Franco de Andrade-Júnior, Camila Aparecida de Carvalho, Josemar Coelho Lima, Hilton César Rodrigues Magalhães
ABSTRAKT
The increased incidence of visceral leishmaniasis (VL) in Brazil is due to a lack of effective disease control measures. In addition to that, no effective treatment exists for canine VL in response to synthetic drugs. Thus, the objective of this study was to evaluate the effect of the essential oils of Coriandrum sativum and Lippia sidoides, and oleoresin from Copaifera reticulata, on Leishmania chagasi promastigotes and amastigotes. We also examined the toxicity of these treatments on the murine monocyte cell line RAW 264.7. To determine the IC50 a MTT test (3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) was performed on promastigotes, and an in situ ELISA assay was conducted on amastigotes. Here, we demonstrate that oleoresin from C. reticulata was effective against both promastigotes (IC50 of 7.88 µg.mL-1) and amastigotes (IC50 of 0.52 µg.mL-1), and neither of the two treatments differed significantly (p > 0.05) from pentamidine (IC50 of 2.149 µg.mL-1) and amphotericin B (IC50 of 9.754 µg.mL-1). Of the three plant oils tested, only oleoresin showed no toxicity toward monocyte, with 78.45% viability after treatment. Inhibition of promastigote and amastigote growth and the lack of cytotoxicity by C. reticulata demonstrate that oleoresin may be a viable option for analyzing the in vivo therapeutic effects of leishmanicidal plants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
RAW 264.7 Cell Line murine, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. 91062702, mouse blood, Macrophage