Merck
  • Simulation-Assisted Determination of the Minimum Melting Temperature Composition of MgCl2?KCl?NaCl Salt Mixture for Next-Generation Molten Salt Thermal Energy Storage
Simulation-Assisted Determination of the Minimum Melting Temperature Composition of MgCl2?KCl?NaCl Salt Mixture for Next-Generation Molten Salt Thermal Energy Storage
Frontiers in energy research (2022)
Villada C, et.al