Merck
  • OCT3/4 is a potential immunohistochemical biomarker for diagnosis and prognosis of primary intracranial germ cell tumors: a systematic review and meta-analysis.
OCT3/4 is a potential immunohistochemical biomarker for diagnosis and prognosis of primary intracranial germ cell tumors: a systematic review and meta-analysis.
Frontiers in neuroscience (2023-07-21)
Yi Zhang, Mucong Li, Jifang Liu, Kan Deng, Huijuan Zhu, Lin Lu, Hui Pan, Renzhi Wang, Yong Yao
ABSTRAKT
Intracranial germ cell tumors (iGCTs), comprising of germinoma (GE) and non-germinomatous GCT (NGGCT), are a group of heterogenous brain tumors. Immunohistochemical markers, such as placental-like alkaline phosphatase (PLAP), are commonly used in diagnosis but show moderate sensitivity. Organic cation transporter 3/4 (OCT3/4) has been proposed as a novel biomarker for diagnosis and prognosis of iGCTs. This paper aimed to compare OCT3/4 with PLAP as potential immunohistochemical biomarkers in iGCTs diagnosis and clarify the relationship between OCT3/4 and prognosis of patients with iGCTs. Meta-analyses were performed to estimate pooled percentage point differences in positive rates between OCT3/4 and PLAP, their sensitivities, and correlation between OCT3/4 and prognosis in iGCTs. Nine articles were included representing of 241 patients. A fixed-effects model meta-analysis revealed that OCT3/4s positive rate was 8.6% higher (95% CI, 0.7% lower to 17.9% higher) than that of PLAP. Using fixed-effects models, sensitivities of OCT3/4 as a potential immunohistochemical biomarker in CNS GE and NGGCT were 85% (95% CI, 79% to 89%) and 56% (95% CI, 39% to 71%), respectively. In comparison, PLAP had lower sensitivities in both GE (73%; 95% CI, 64% to 91%) and NGGCT (43%; 95% CI, 27% to 61%). Moreover, OCT3/4 was significantly negatively correlated with 5-year progression free survival in patients with CNS GE (HR = 2.56, 95 % CI 1.47 to 4.44; p = 0.0008). Sensitivity analyses showed similar results. This study provides the first comprehensive assessment of the efficacies of OCT3/4 and PLAP in iGCTs detection and prognosis prediction, indicating OCT3/4 seems to be a more sensitive and reliable immunohistochemical marker in iGCT diagnosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Oct-4 Antibody, Chemicon®, from rabbit