Merck
Mitochondrial Genome-Encoded Long Noncoding RNA and Mitochondrial Stability in Diabetic Retinopathy.
Diabetes (2022-12-24)
Jay Kumar, Ghulam Mohammad, Kumari Alka, Renu A Kowluru
ABSTRAKT
Mitochondria experience genomic and functional instability in diabetes, and mitochondrial dysfunction has a critical role in the development of diabetic retinopathy. Diabetes also alters expressions of many long noncoding RNAs (LncRNAs), the RNAs with >200 nucleotides and no open reading frame. LncRNAs are mainly encoded by the nuclear genome, but mtDNA also encodes three LncRNAs. Our goal was to investigate the effect of hyperglycemia on mtDNA-encoded LncRNA cytochrome B (LncCytB) in mtDNA stability in diabetic retinopathy. Retinal endothelial cells, transfected with LncCytB-overexpressing plasmids or siRNA, incubated in 5 mmol/L d-glucose (normal glucose [NG]) or 20 mmol/L d-glucose (high glucose [HG]) for 4 days, were analyzed for LncCytB expression by strand-specific PCR and its mitochondrial localization by RNA fluorescence in situ hybridization. Damage-sensitive mtDNA regions were examined by micrococcal nuclease (MNase) digestion sequencing and LncCytB occupancy at mtDNA by chromatin isolation by RNA purification. Protective nucleoids in mtDNA were analyzed by SYBR Green-MitoTracker Red staining and confirmed in isolated mitochondria by flow cytometry. Compared with NG, HG downregulated LncCytB by >50% but had no significant effect on the other mtDNA-encoded LncRNAs. mtDNA packaging was impaired, MNase sensitivity was increased, and LncCytB occupancy at mtDNA was decreased. While LncCytB overexpression ameliorated mtDNA damage and decrease in nucleoids and copy numbers, LncCytB-siRNA exacerbated damage and further reduced nucleoids. Retinal microvessels from streptozotocin-induced diabetic mice and human donors with diabetic retinopathy presented a similar decrease in LncCytB and mtDNA nucleoids. Thus, LncCytB has a major role in maintaining mitochondrial genomic stability, and its downregulation in the hyperglycemic milieu contributes to increased vulnerability of mtDNA to damage.