Merck
Characterization of KLHL14 anti-oncogenic action in malignant mesothelioma.
Heliyon (2024-03-21)
Angelo Canciello, Reyes Benot Domínguez, Barbara Barboni, Antonio Giordano, Andrea Morrione
ABSTRAKT
Malignant mesothelioma (MM) is a very aggressive neoplasia with a short life expectancy and limited therapeutic options. Thus, the identification of novel molecular targets is a matter of great urgency. Kelch-like (KLHL) proteins play an important role in a number of physiological and pathological cell-regulatory processes. Among this family, the function of KLHL14 is still very poorly characterized. KLHL14 was originally identified as a gene involved in regulating the epithelial-mesenchymal transition (EMT) process. Here, we demonstrate that KLHL14 not only prevents EMT but also plays an anti-oncogenic role in MM. Indeed, KLHL14 depletion enhanced proliferation, motility, invasion and colony formation in MM cells. Importantly, we also demonstrated that KLHL14 mechanism of action is dependent on Transforming Growth Factor β (TGF-β). In fact, TGF-β promotes de novo synthesis, increases protein stability and induces nuclear-cytoplasmic shuttling of KLHL14. Collectively, this research is an important step further to decipher KLHLs mechanism of action and further contributes to the understanding of the molecular mechanisms regulating MM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture