Merck
Macrophage-coated tumor cluster aggravates hepatoma invasion and immunotherapy resistance via generating local immune deprivation.
Cell reports. Medicine (2024-04-14)
Junya Ning, Yingnan Ye, Hongru Shen, Runjiao Zhang, Huikai Li, Tianqiang Song, Rui Zhang, Pengpeng Liu, Guidong Chen, Hailong Wang, Fenglin Zang, Xiangchun Li, Jinpu Yu
ABSTRAKT
Immune checkpoint inhibitors (ICIs) represent a promising treatment for hepatocellular carcinoma (HCC) due to their capacity for abundant lymphocyte infiltration. However, some patients with HCC respond poorly to ICI therapy due to the presence of various immunosuppressive factors in the tumor microenvironment. Our research reveals that a macrophage-coated tumor cluster (MCTC) signifies a unique spatial structural organization in HCC correlating with diminished recurrence-free survival and overall survival in a total of 572 HCC cases from 3 internal cohorts and 2 independent external validation cohorts. Mechanistically, tumor-derived macrophage-associated lectin Mac-2 binding protein (M2BP) induces MCTC formation and traps immunocompetent cells at the edge of MCTCs to induce intratumoral cytotoxic T cell exclusion and local immune deprivation. Blocking M2BP with a Mac-2 antagonist might provide an effective approach to prevent MCTC formation, enhance T cell infiltration, and thereby improve the efficacy of ICI therapy in HCC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid